Assay Details
Target Gene Details
Entrez Gene ID: | 14874 |
Gene Name: | glutathione transferase zeta 1 (maleylacetoacetate isomerase) |
Gene Aliases: |
MAAI |
Location: |
Chr.12:87147165-87164723 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstz1 | NM_001252555.1 | NP_001239484.1 | ||
| NM_001252556.1 | NP_001239485.1 | |||
| NM_010363.4 | NP_034493.1 | |||
| AB041613.1 | BAA95096.1 | |||
| AF093418.1 | AAD43846.1 | |||
| AK002398.1 | BAB22070.1 | |||
| AK012796.1 | ||||
| AK075927.1 | BAC36059.1 | |||
| BC031777.1 | AAH31777.1 | |||
| BF722376.1 |
More Information
Set Membership: |
|

Genomic Map