Assay Details
Target Gene Details
Entrez Gene ID: | 217882 |
Gene Name: | centrosomal protein 170B |
Gene Aliases: |
AI466840, AW555464, Kiaa0284, mKIAA0284 |
Location: |
Chr.12:112720591-112746592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep170b | NM_001024602.3 | NP_001019773.2 | ||
| AK122236.1 | BAC65518.1 | |||
| AK147410.1 | BAE27896.1 | |||
| AK147612.1 | BAE28024.1 | |||
| BC050077.1 | AAH50077.1 | |||
| BC141174.1 |
More Information
Set Membership: |
|

Genomic Map