Assay Details
Target Gene Details
Entrez Gene ID: | 56784 |
Gene Name: | Ral GTPase activating protein, alpha subunit 1 |
Gene Aliases: |
2310003F20Rik, 4930400K19Rik, AI563624, GRIPE, Garnl1, Tulip1, mKIAA0884 |
Location: |
Chr.12:55602890-55821575 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgapa1 | NM_001003719.2 | 22 | 3945 | NP_001003719.1 |
| NM_001112714.2 | 22 | 3945 | NP_001106185.1 | |
| NM_001286263.1 | 23 | 4086 | NP_001273192.1 | |
| NM_019994.5 | 22 | 3945 | NP_064378.4 | |
| AY066011.1 | 10 | 1531 | AAL47577.1 | |
| AY596972.1 | 22 | 3596 | AAT49273.1 | |
| AY596973.1 | 22 | 3596 | AAT49274.1 | |
| AY596974.1 | 22 | 3596 | AAT49275.1 | |
| BC145119.1 | 23 | 3667 | ||
| BC150707.1 | 23 | 3667 |
More Information
Set Membership: |
|

Genomic Map