Assay Details
Target Gene Details
Entrez Gene ID: | 19179 |
Gene Name: | protease (prosome, macropain) 26S subunit, ATPase 1 |
Gene Aliases: |
AI325227, P26s4, Rpt2/S4, S4, rpt2 |
Location: |
Chr.12:100110202-100123364 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc1 | NM_008947.3 | NP_032973.1 | ||
| AK088411.1 | BAC40339.1 | |||
| AK133926.1 | BAE21929.1 | |||
| AK148906.1 | BAE28695.1 | |||
| AK161641.1 | BAE36506.1 | |||
| AK161723.1 | ||||
| AK166783.1 | BAE39016.1 | |||
| AK169015.1 | BAE40812.1 | |||
| BC003860.1 | AAH03860.1 | |||
| U39302.1 | AAB34137.1 |
More Information
Set Membership: |
|

Genomic Map