Assay Details
Target Gene Details
Entrez Gene ID: | 66695 |
Gene Name: | asporin |
Gene Aliases: |
4631401G09Rik, AA986886, Plap1, Slrr1c |
Location: |
Chr.13:49544443-49567565 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aspn | NM_001172481.1 | NP_001165952.1 | ||
| NM_025711.3 | NP_079987.2 | |||
| AF316825.1 | AAK35162.1 | |||
| AK014504.1 | ||||
| AK021386.1 | BAB32393.1 | |||
| AK085920.1 | ||||
| BC034888.1 | AAH34888.1 | |||
| BC145904.1 | ||||
| BC145930.1 |
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpp | NM_025495.3 | NP_079771.1 | ||
| AK006234.1 | BAB24472.1 | |||
| AK012863.1 | BAB28519.1 | |||
| AK014919.1 | BAB29620.1 | |||
| BC147207.1 | ||||
| BC147208.1 |
More Information
Set Membership: |
|

Genomic Map