Assay Details
Target Gene Details
Entrez Gene ID: | 24000 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 21 |
Gene Aliases: |
PTPD1, PTPRL10 |
Location: |
Chr.12:98676741-98737618 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn21 | NM_001146199.1 | NP_001139671.1 | ||
| NM_011877.2 | NP_036007.2 | |||
| BC014691.1 | ||||
| BC066856.1 | AAH66856.1 | |||
| BC098500.1 | AAH98500.1 | |||
| D37801.1 | BAA07053.1 |
More Information
Set Membership: |
|

Genomic Map