Assay Details
Target Gene Details
Entrez Gene ID: | 110417 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class H |
Gene Aliases: |
2210416H01Rik, A930028P05Rik |
Location: |
Chr.12:79080664-79089754 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigh | NM_029988.2 | 2 | 377 | NP_084264.1 |
| AK020902.1 | 2 | 353 | ||
| AK030744.1 | 2 | 341 | BAC27112.1 | |
| AK167892.1 | 2 | 359 | BAE39904.1 | |
| BC086806.1 | 2 | 328 | AAH86806.1 | |
| BY737882.1 | 2 | 359 | ||
| BY779412.1 |
More Information
Set Membership: |
|

Genomic Map