Assay Details
Target Gene Details
Entrez Gene ID: | 26931 |
Gene Name: | protein phosphatase 2, regulatory subunit B', gamma |
Gene Aliases: |
2610043M05Rik, 2700063L20Rik, AI060890, AW545884, C85228, D12Bwg0916e, mKIAA0044 |
Location: |
Chr.12:110447179-110583075 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5c | NM_001135001.1 | NP_001128473.1 | ||
| AK129042.1 | BAC97852.1 | |||
| AK161855.1 | BAE36607.1 | |||
| CX217314.1 |
More Information
Set Membership: |
|

Genomic Map