Assay Details
Target Gene Details
Entrez Gene ID: | 432713 |
Gene Name: | predicted gene 5441 |
Gene Aliases: |
- |
Location: |
Chr.12:117234521-117337012 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5441 | NR_044987.1 |
Target Gene Details
Entrez Gene ID: | 19276 |
Gene Name: | protein tyrosine phosphatase, receptor type, N polypeptide 2 |
Gene Aliases: |
4930425H11Rik, IA2beta, PTP-NP, Phol, mKIAA0387, phogrin |
Location: |
Chr.12:116485692-117278167 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprn2 | NM_011215.2 | NP_035345.2 | ||
| AK138655.1 | BAE23734.1 | |||
| BC133678.1 | ||||
| U57345.1 | AAB06945.1 | |||
| U82439.1 | AAB39996.1 |
More Information
Set Membership: |
|

Genomic Map