Assay Details
Target Gene Details
Entrez Gene ID: | 19276 |
Gene Name: | protein tyrosine phosphatase, receptor type, N polypeptide 2 |
Gene Aliases: |
4930425H11Rik, IA2beta, PTP-NP, Phol, mKIAA0387, phogrin |
Location: |
Chr.12:116485692-117278167 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprn2 | NM_011215.2 | 5 | 644 | NP_035345.2 |
| AK081267.1 | 5 | 636 | ||
| AK083776.1 | 5 | 595 | ||
| AK138655.1 | 5 | 636 | BAE23734.1 | |
| AK220255.1 | 5 | 572 | BAD90180.1 | |
| BC133678.1 | 5 | 528 | ||
| U57345.1 | 5 | 531 | AAB06945.1 |
More Information
Set Membership: |
|

Genomic Map