Assay Details
Target Gene Details
Entrez Gene ID: | 50779 |
Gene Name: | regulator of G-protein signaling 6 |
Gene Aliases: |
- |
Location: |
Chr.12:82616176-83158815 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs6 | NM_001282061.1 | NP_001268990.1 | ||
NM_015812.3 | NP_056627.1 | |||
AF061933.1 | AAC70011.1 | |||
AK052527.1 | BAC35026.1 | |||
AK053087.1 | BAC35262.1 | |||
BC125029.1 | ||||
BC125030.1 |
