Assay Details
Target Gene Details
Entrez Gene ID: | 26932 |
Gene Name: | protein phosphatase 2, regulatory subunit B', epsilon |
Gene Aliases: |
4633401M22Rik, AI449017, B56beta |
Location: |
Chr.12:75449463-75596250 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5e | NM_012024.2 | NP_036154.1 | ||
| AK053228.1 | ||||
| AK088366.1 | BAC40306.1 | |||
| AK132432.1 | BAE21166.1 | |||
| AK133824.1 | BAE21863.1 | |||
| AK220163.1 | BAD90335.1 | |||
| BC019199.1 | ||||
| BC085149.1 | AAH85149.1 | |||
| U49728.1 | AAB37234.1 |
More Information
Set Membership: |
|

Genomic Map