Assay Details
Target Gene Details
Entrez Gene ID: | 72736 |
Gene Name: | thioredoxin-related transmembrane protein 1 |
Gene Aliases: |
2810425A04Rik, AI465319, Txndc1 |
Location: |
Chr.12:70453154-70467624 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmx1 | NM_028339.1 | NP_082615.1 | ||
| AK013150.1 | BAB28680.3 | |||
| AK030429.1 | BAC26960.1 | |||
| AK050567.1 | BAC34326.1 | |||
| AK137059.1 | BAE23223.1 | |||
| AK150511.1 | BAE29624.1 | |||
| AK153027.1 | BAE31660.1 | |||
| AK170468.1 | BAE41816.1 | |||
| BC017603.1 | AAH17603.1 | |||
| BC021533.1 | AAH21533.1 |
More Information
Set Membership: |
|

Genomic Map