Assay Details
Target Gene Details
Entrez Gene ID: | 218214 |
Gene Name: | lysine (K)-specific demethylase 1B |
Gene Aliases: |
4632428N09Rik, AI482520, Aof1 |
Location: |
Chr.13:47043471-47085279 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm1b | NM_172262.3 | 15 | 1935 | NP_758466.1 |
| AK028553.1 | 5 | 601 | BAC26005.1 | |
| AK078920.1 | 4 | 493 | BAC37460.1 | |
| BC023917.1 | 15 | 1876 | AAH23917.1 | |
| BY714054.1 |
More Information
Set Membership: |
|

Genomic Map