Assay Details
Target Gene Details
Entrez Gene ID: | 104871 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
AI661438, B230306G18Rik, HSD3 |
Location: |
Chr.12:98628123-98669819 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata7 | NM_001289572.1 | NP_001276501.1 | ||
| NM_001289573.1 | NP_001276502.1 | |||
| NM_001289574.1 | NP_001276503.1 | |||
| NM_178914.4 | NP_849245.2 | |||
| AK036359.1 | ||||
| AK077500.1 | BAC36832.1 | |||
| AK153698.1 | BAE32149.1 | |||
| AK161308.1 | BAE36312.1 | |||
| BC046960.1 | AAH46960.1 | |||
| DQ196101.1 |
More Information
Set Membership: |
|

Genomic Map