Assay Details
Target Gene Details
Entrez Gene ID: | 320244 |
Gene Name: | tubulin tyrosine ligase-like family, member 5 |
Gene Aliases: |
1700048H13Rik, 2310009M18Rik, 4930556H18Rik, AI428864, D630041K24Rik, R75373, Stamp, mKIAA0998 |
Location: |
Chr.12:85824950-86053760 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll5 | NM_001081423.2 | NP_001074892.2 | ||
| AB093278.1 | BAC41462.1 | |||
| AK029125.1 | BAC26310.1 | |||
| AK085566.1 | BAC39474.1 | |||
| AM690748.1 | ||||
| BC035276.1 | AAH35276.1 |
More Information
Set Membership: |
|

Genomic Map