Assay Details
Target Gene Details
Entrez Gene ID: | 76251 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 |
Gene Aliases: |
0610007P08Rik, 1700019D06Rik, 9330134C04Rik, Rad26l, Sr278 |
Location: |
Chr.13:63813415-63900302 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc6l2 | NM_001013608.2 | NP_001013626.2 | ||
NM_023507.3 | NP_075996.2 | |||
AF217319.1 | AAF73858.1 | |||
AK035054.1 | BAC28927.1 | |||
AK049874.1 | ||||
AK082850.1 | BAC38652.1 | |||
BC032964.1 | AAH32964.1 | |||
CN457854.1 | ||||
CO430774.1 |
