Assay Details
Target Gene Details
Entrez Gene ID: | 104871 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
AI661438, B230306G18Rik, HSD3 |
Location: |
Chr.12:98628123-98669819 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spata7 | NM_001289572.1 | 7 | 1008 | NP_001276501.1 |
NM_001289573.1 | 6 | 603 | NP_001276502.1 | |
NM_001289574.1 | 8 | 1170 | NP_001276503.1 | |
NM_178914.4 | 8 | 1104 | NP_849245.2 | |
AK036359.1 | 8 | 1088 | ||
AK077500.1 | 8 | 1015 | BAC36832.1 | |
AK153698.1 | 6 | 579 | BAE32149.1 | |
AK161308.1 | 7 | 954 | BAE36312.1 | |
BC046960.1 | 8 | 1062 | AAH46960.1 | |
DQ196101.1 | 7 | 834 |
