Assay Details
Target Gene Details
Entrez Gene ID: | 76251 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 |
Gene Aliases: |
0610007P08Rik, 1700019D06Rik, 9330134C04Rik, Rad26l, Sr278 |
Location: |
Chr.13:63813415-63900302 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc6l2 | NM_001013608.2 | 4 | 797 | NP_001013626.2 |
NM_023507.3 | 4 | 783 | NP_075996.2 | |
AF217319.1 | 3 | 600 | AAF73858.1 | |
AK002307.1 | 4 | 784 | BAB22002.1 | |
AK035054.1 | 4 | 749 | BAC28927.1 | |
AK049874.1 | 4 | 1109 | ||
AK082850.1 | 3 | 452 | BAC38652.1 | |
BU148612.1 | 4 | 815 |
