Assay Details
Target Gene Details
Entrez Gene ID: | 74069 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3A |
Gene Aliases: |
- |
Location: |
Chr.12:104106704-104121897 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina3a | NM_001167705.1 | NP_001161177.1 | ||
| NM_028740.2 | NP_083016.1 | |||
| AK016702.1 | BAB30388.1 | |||
| BC063325.1 | AAH63325.1 |
More Information
Set Membership: |
|

Genomic Map