Assay Details
Target Gene Details
Entrez Gene ID: | 22031 |
Gene Name: | TNF receptor-associated factor 3 |
Gene Aliases: |
AI528849, CAP-1, CD40bp, CRAF1, LAP1, T-BAM, amn |
Location: |
Chr.12:111166370-111267155 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf3 | NM_001286122.1 | NP_001273051.1 | ||
| NM_011632.3 | NP_035762.2 | |||
| AK147304.1 | BAE27835.1 | |||
| AK147362.1 | BAE27866.1 | |||
| AK148231.1 | ||||
| BC137634.1 | ||||
| BC137635.1 | ||||
| U21050.1 | AAC52175.1 |
More Information
Set Membership: |
|

Genomic Map