Assay Details
Target Gene Details
Entrez Gene ID: | 217843 |
Gene Name: | unc-79 homolog (C. elegans) |
Gene Aliases: |
9030205A07Rik, Mlca3 |
Location: |
Chr.12:102948329-103184228 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc79 | NM_001081017.2 | 20 | 3429 | NP_001074486.2 |
| AB093296.1 | 3 | 517 | BAD02453.1 | |
| AB257853.1 | 20 | 3429 | ||
| GQ334471.1 | 20 | 2575 |

Genomic Map