Assay Details
Target Gene Details
Entrez Gene ID: | 211945 |
Gene Name: | pleckstrin homology domain containing, family H (with MyTH4 domain) member 1 |
Gene Aliases: |
AV308913, C77772, D630024D12Rik, mKIAA1200 |
Location: |
Chr.12:79029163-79081655 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhh1 | NM_181073.3 | NP_851418.2 | ||
| AK122464.1 | BAC65746.1 | |||
| AK143387.1 | BAE25361.1 |

Genomic Map