Assay Details
Target Gene Details
Entrez Gene ID: | 217843 |
Gene Name: | unc-79 homolog (C. elegans) |
Gene Aliases: |
9030205A07Rik, Mlca3 |
Location: |
Chr.12:102948329-103184228 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc79 | NM_001081017.2 | 28 | 4823 | NP_001074486.2 |
AB093296.1 | 11 | 1911 | BAD02453.1 | |
AB257853.1 | 28 | 4823 | ||
GQ334471.1 | 29 | 4035 |
