Assay Details
Target Gene Details
Entrez Gene ID: | 68675 |
Gene Name: | family with sequence similarity 172, member A |
Gene Aliases: |
1110033M05Rik, 2610318O14Rik, 53-E6, 9430037D06Rik, AF064782, pEN87 |
Location: |
Chr.13:77708678-78166240 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam172a | NM_001163419.1 | NP_001156891.1 | ||
| NM_001163420.1 | NP_001156892.1 | |||
| NM_138312.1 | NP_612185.1 | |||
| NR_028109.1 | ||||
| AF064782.1 | AAD15841.1 | |||
| AK052419.1 | ||||
| AK165273.1 | BAE38114.1 | |||
| AK166285.1 | BAE38682.1 | |||
| BC057380.1 | AAH57380.1 |

Genomic Map