Assay Details
Target Gene Details
Entrez Gene ID: | 18760 |
Gene Name: | protein kinase D1 |
Gene Aliases: |
PKD, PKD1, Pkcm, Prkcm |
Location: |
Chr.12:50341226-50649248 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkd1 | NM_008858.3 | 6 | 1076 | NP_032884.2 |
| Z34524.1 | 6 | 951 | CAA84283.1 |
More Information
Set Membership: |
|

Genomic Map