Assay Details
Target Gene Details
Entrez Gene ID: | 104886 |
Gene Name: | RAB15, member RAS oncogene family |
Gene Aliases: |
2310012G06Rik, AI840042 |
Location: |
Chr.12:76797960-76822761 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab15 | NM_134050.4 | NP_598811.3 | ||
AB232610.1 | BAF02872.1 | |||
AK158637.1 | BAE34591.1 | |||
AK158768.1 | BAE34652.1 | |||
BC013790.1 | AAH13790.1 | |||
BC027769.1 | AAH27769.1 |
