Assay Details
Target Gene Details
Entrez Gene ID: | 20384 |
Gene Name: | serine/arginine-rich splicing factor 5 |
Gene Aliases: |
Sfrs5 |
Location: |
Chr.12:80945504-80950507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf5 | NM_001079694.1 | NP_001073162.1 | ||
| NM_001079695.1 | NP_001073163.1 | |||
| NM_009159.2 | NP_033185.2 | |||
| AK002267.1 | ||||
| AK007729.1 | BAB25217.1 | |||
| AK078065.1 | ||||
| AK088358.1 | ||||
| AK142009.1 | ||||
| AK146624.1 | BAE27313.1 | |||
| BC082593.1 | AAH82593.1 | |||
| BC085267.1 | AAH85267.1 | |||
| BU935699.1 |
More Information
Set Membership: |
|

Genomic Map