Assay Details
Target Gene Details
Entrez Gene ID: | 94186 |
Gene Name: | striatin, calmodulin binding protein 3 |
Gene Aliases: |
Gs2na, SG2NA |
Location: |
Chr.12:51608541-51692565 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Strn3 | NM_001172098.1 | NP_001165569.1 | ||
NM_052973.2 | NP_443205.1 | |||
AF307777.1 | AAG24454.1 | |||
AK140447.1 | ||||
AK165908.1 | ||||
BC138055.1 | ||||
BC145127.1 | ||||
EF685152.1 |
