Assay Details
Target Gene Details
Entrez Gene ID: | 76357 |
Gene Name: | TRM5 tRNA methyltransferase 5 |
Gene Aliases: |
2610027O18Rik, mKIAA1393 |
Location: |
Chr.12:73268515-73286787 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt5 | NM_029580.3 | 5 | 1516 | NP_083856.1 |
| AK011576.1 | 5 | 1517 | BAB27710.1 | |
| AK049127.1 | 5 | 1515 | ||
| AK220389.1 | 4 | 1312 | BAD90443.1 | |
| BC012521.1 | 5 | 1517 | AAH12521.1 |
More Information
Set Membership: |
|

Genomic Map