Assay Details
Target Gene Details
Entrez Gene ID: | 218639 |
Gene Name: | ADP-ribosylation factor-like 15 |
Gene Aliases: |
A430036I03, Arfrp2, C230032K13Rik |
Location: |
Chr.13:113794506-114157462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl15 | NM_172595.4 | NP_766183.1 | ||
AK039965.1 | BAC30485.1 | |||
AK048736.1 | BAC33440.1 | |||
AK050979.1 | BAE20688.1 | |||
AK052164.1 | BAC34864.1 | |||
AK053953.1 | ||||
BC120664.1 | ||||
BC120666.1 | ||||
BC144993.1 | ||||
CJ063085.1 | ||||
CJ066332.1 |
