Assay Details
Target Gene Details
Entrez Gene ID: | 18976 |
Gene Name: | pro-opiomelanocortin-alpha |
Gene Aliases: |
ACTH, BE, Beta-LPH, Clip, Gamma-LPH, Npp, Pomc-1, Pomc1, alpha-MSH, alphaMSH, beta-MSH, gamma-MSH |
Location: |
Chr.12:3954945-3960643 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomc | NM_001278581.1 | NP_001265510.1 | ||
| NM_001278582.1 | NP_001265511.1 | |||
| NM_001278583.1 | NP_001265512.1 | |||
| NM_001278584.1 | NP_001265513.1 | |||
| NM_008895.4 | NP_032921.1 | |||
| AK017492.1 | BAB30771.1 | |||
| AK017581.1 | BAB30818.1 | |||
| AK030714.1 | BAC27095.1 | |||
| AK077426.1 | BAC36795.1 | |||
| AK133592.1 | BAE21739.1 | |||
| AK133646.1 | BAE21764.1 | |||
| AK133740.1 | BAE21815.1 | |||
| AK133776.1 | BAE21833.1 | |||
| AK133800.1 | BAE21850.1 | |||
| BC061215.1 | AAH61215.1 |

Genomic Map