Assay Details
Target Gene Details
Entrez Gene ID: | 217734 |
Gene Name: | protein-O-mannosyltransferase 2 |
Gene Aliases: |
A830009D15Rik, AW046274 |
Location: |
Chr.12:87106861-87147902 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt2 | NM_153415.3 | NP_700464.2 | ||
| AF246235.1 | AAL85627.1 | |||
| AK043573.1 | ||||
| AK043713.1 | ||||
| AK050915.1 | BAC34458.1 | |||
| AK076813.1 | ||||
| AK214879.1 | ||||
| AY090481.1 | AAM12047.1 | |||
| AY090482.1 | AAM12048.1 | |||
| BC052045.1 | AAH52045.1 | |||
| BC062965.1 | AAH62965.1 |
More Information
Set Membership: |
|

Genomic Map