Assay Details
Target Gene Details
Entrez Gene ID: | 13612 |
Gene Name: | EGF-like repeats and discoidin I-like domains 3 |
Gene Aliases: |
Del-1, Del1 |
Location: |
Chr.13:88821472-89323225 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Edil3 | NM_001037987.3 | 2 | 641 | NP_001033076.1 |
| NM_010103.4 | 2 | 641 | NP_034233.1 | |
| AF031524.1 | 2 | 749 | AAB86585.1 | |
| AF031525.1 | 2 | 680 | AAB86586.1 | |
| AK036117.1 | 2 | 602 | BAC29310.1 | |
| AK044978.1 | 2 | 500 | ||
| AK045088.1 | 2 | 457 | ||
| AK045839.1 | 2 | 558 | BAC32509.1 | |
| AK046885.1 | 2 | 642 | BAC32909.1 | |
| AK081003.1 | 2 | 473 | BAC38111.1 | |
| AK084215.1 | 2 | 496 | ||
| AK135220.1 | 2 | 605 | ||
| AK139093.1 | 2 | 603 | BAE23887.1 | |
| AK141907.1 | 2 | 625 | ||
| AK157810.1 | 2 | 626 | ||
| BC056386.1 | 2 | 541 | AAH56386.1 |
More Information
Set Membership: |
|

Genomic Map