Assay Details
Target Gene Details
Entrez Gene ID: | 13612 |
Gene Name: | EGF-like repeats and discoidin I-like domains 3 |
Gene Aliases: |
Del-1, Del1 |
Location: |
Chr.13:88821472-89323225 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Edil3 | NM_001037987.3 | 4 | 797 | NP_001033076.1 |
| NM_010103.4 | 3 | 767 | NP_034233.1 | |
| AF031524.1 | 4 | 905 | AAB86585.1 | |
| AF031525.1 | 4 | 836 | AAB86586.1 | |
| AK036117.1 | 4 | 758 | BAC29310.1 | |
| AK044978.1 | 3 | 626 | ||
| AK045088.1 | 3 | 583 | ||
| AK046885.1 | 4 | 798 | BAC32909.1 | |
| AK081003.1 | 3 | 599 | BAC38111.1 | |
| AK084215.1 | 4 | 652 | ||
| AK135220.1 | 4 | 761 | ||
| AK139093.1 | 4 | 759 | BAE23887.1 | |
| AK141907.1 | 4 | 781 | ||
| AK157810.1 | 4 | 782 | ||
| BC056386.1 | 4 | 697 | AAH56386.1 |
More Information
Set Membership: |
|

Genomic Map