Assay Details
Target Gene Details
Entrez Gene ID: | 68083 |
Gene Name: | PAK1 interacting protein 1 |
Gene Aliases: |
5830431I15Rik, 5930415H02Rik, AA419825, AI314040, AW556169, Gdpd1, PIP1 |
Location: |
Chr.13:41001010-41013033 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak1ip1 | NM_026550.3 | NP_080826.2 | ||
| AF386076.1 | AAL40652.1 | |||
| AK002852.1 | BAB22407.1 | |||
| AK017959.1 | 3 | 643 | ||
| AK077846.1 | ||||
| AK077941.1 | BAC37076.1 | |||
| AK135946.1 | BAE22737.1 | |||
| AK150747.1 | BAE29818.1 | |||
| AK150919.1 | BAE29957.1 | |||
| AK188749.1 | ||||
| AK189086.1 | ||||
| AY030406.1 | AAK51599.1 |
More Information
Set Membership: |
|

Genomic Map