Assay Details
Target Gene Details
Entrez Gene ID: | 218194 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
9630030F18Rik, Rpel1 |
Location: |
Chr.13:42680571-43138526 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr1 | NM_001005740.1 | 7 | 1323 | NP_001005740.1 |
| AK048208.1 | 7 | 1219 | BAC33272.1 | |
| AK139452.1 | 7 | 1295 | ||
| AY993932.1 | 8 | 1260 | AAY42814.1 | |
| BC048407.1 | 6 | 1154 | AAH48407.1 | |
| BC061691.1 | 7 | 1323 | AAH61691.1 |
More Information
Set Membership: |
|

Genomic Map