Assay Details
Target Gene Details
Entrez Gene ID: | 218194 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
9630030F18Rik, Rpel1 |
Location: |
Chr.13:42680571-43138526 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr1 | NM_001005740.1 | NP_001005740.1 | ||
| NM_001005748.2 | NP_001005748.1 | |||
| AK048208.1 | BAC33272.1 | |||
| AK139452.1 | ||||
| AK166463.1 | BAE38791.1 | |||
| AY993932.1 | AAY42814.1 | |||
| BC048407.1 | AAH48407.1 | |||
| BC048620.1 | AAH48620.1 | |||
| BC061691.1 | AAH61691.1 | |||
| CD551063.1 |
More Information
Set Membership: |
|

Genomic Map