Assay Details
Target Gene Details
Entrez Gene ID: | 12572 |
Gene Name: | cyclin-dependent kinase 7 |
Gene Aliases: |
AI323415, AI528512, C230069N13, Cdkn7, Crk4 |
Location: |
Chr.13:100697024-100730942 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk7 | NM_009874.3 | NP_034004.2 | ||
| AK039084.1 | BAC30233.1 | |||
| AK145759.1 | BAE26631.1 | |||
| AK168286.1 | BAE40231.1 | |||
| BC004605.1 | AAH04605.1 | |||
| BC068160.1 | AAH68160.1 | |||
| BC141043.1 | ||||
| U11822.1 | AAA64831.1 | |||
| X74145.1 | CAA52242.1 |
More Information
Set Membership: |
|

Genomic Map