Assay Details
Target Gene Details
Entrez Gene ID: | 238875 |
Gene Name: | Grb2-binding adaptor, transmembrane |
Gene Aliases: |
9830130M13Rik |
Location: |
Chr.13:110352614-110357273 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gapt | NM_177713.3 | NP_808381.1 | ||
| AK036534.1 | BAC29466.1 | |||
| AK155882.1 | BAE33480.1 |

Genomic Map