Assay Details
Target Gene Details
Entrez Gene ID: | 13361 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
8430436I03Rik, AA607882, AI662710, AW555094 |
Location: |
Chr.13:92354783-92389053 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhfr | NM_010049.3 | 1 | 60 | NP_034179.1 |
| AK136212.1 | 1 | 117 | BAE22879.1 | |
| BU058996.1 | 1 | 60 |
Target Gene Details
Entrez Gene ID: | 17686 |
Gene Name: | mutS homolog 3 (E. coli) |
Gene Aliases: |
D13Em1, Rep-3, Rep3 |
Location: |
Chr.13:92211879-92355003 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh3 | NM_010829.2 | 1 | 162 | NP_034959.2 |
| AK011426.1 | 1 | 145 | ||
| AK040616.1 | 1 | 156 | BAC30646.1 | |
| AK078467.1 | 1 | 113 | BAE43365.1 | |
| AK083388.1 | 1 | 158 | ||
| AK133907.1 | 1 | 155 | ||
| AK161089.1 | 1 | 156 | ||
| BC150759.1 | 1 | 81 | ||
| BY710130.1 | 1 | 144 | ||
| M80360.1 | 1 | 162 | AAA40052.1 |
More Information
Set Membership: |
|

Genomic Map