Assay Details
Target Gene Details
Entrez Gene ID: | 94253 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
9330116H24Rik, AV273951, E130207I19Rik, Nedl1 |
Location: |
Chr.13:14226438-14523280 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hecw1 | NM_001081348.3 | 8 | 1313 | NP_001074817.3 |
| AB083710.1 | 6 | 816 | BAB97389.1 | |
| AK047678.1 | 8 | 1248 | BAC33122.1 | |
| AK051569.1 | 7 | 1134 | BAC34677.1 | |
| AK053694.1 | 7 | 1258 | BAC35477.1 |
More Information
Set Membership: |
|

Genomic Map