Assay Details
Target Gene Details
Entrez Gene ID: | 17686 |
Gene Name: | mutS homolog 3 (E. coli) |
Gene Aliases: |
D13Em1, Rep-3, Rep3 |
Location: |
Chr.13:92211879-92355003 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh3 | NM_010829.2 | 23 | 3200 | NP_034959.2 |
| AK083388.1 | 24 | 3513 | ||
| AK133907.1 | 23 | 3192 | ||
| AK161089.1 | 23 | 3196 | ||
| AK161430.1 | 5 | 618 | BAE36392.1 | |
| BC040784.1 | 8 | 1051 | AAH40784.1 | |
| BC150759.1 | 23 | 3119 | ||
| M80360.1 | 23 | 3185 | AAA40052.1 |
More Information
Set Membership: |
|

Genomic Map