Assay Details
Target Gene Details
Entrez Gene ID: | 54326 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 2 |
Gene Aliases: |
AI317360, Ssc2 |
Location: |
Chr.13:41182382-41220403 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl2 | NM_019423.2 | NP_062296.1 | ||
| AF170908.1 | AAF72573.1 | |||
| AK014803.1 | BAB29559.1 | |||
| AK029855.1 | BAC26646.1 | |||
| AK044770.1 | BAC32079.1 | |||
| AK050397.1 | BAC34236.1 | |||
| BC098215.1 | AAH98215.1 | |||
| BY126714.1 |
More Information
Set Membership: |
|

Genomic Map