Assay Details
Target Gene Details
Entrez Gene ID: | 78689 |
Gene Name: | N(alpha)-acetyltransferase 35, NatC auxiliary subunit |
Gene Aliases: |
A330021G12Rik, A330027C19Rik, AI158944, C030004C14Rik, Mak10 |
Location: |
Chr.13:59585323-59634798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa35 | NM_030153.2 | NP_084429.2 | ||
| AK021042.1 | ||||
| AY102701.1 | AAM52342.1 | |||
| BC027201.1 | ||||
| BC056435.1 | AAH56435.1 |
More Information
Set Membership: |
|

Genomic Map