Assay Details
Target Gene Details
Entrez Gene ID: | 544963 |
Gene Name: | IQ motif containing GTPase activating protein 2 |
Gene Aliases: |
4933417J23Rik, A630053O10, AI788777 |
Location: |
Chr.13:95627177-95891969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqgap2 | NM_027711.1 | NP_081987.1 | ||
| AK147360.1 | ||||
| AK160931.1 | BAE36098.1 | |||
| AK171697.1 | ||||
| AK172491.1 | ||||
| BC042790.1 | AAH42790.1 | |||
| BC150958.1 |
More Information
Set Membership: |
|

Genomic Map