Assay Details
Target Gene Details
Entrez Gene ID: | 66410 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
2410017I18Rik, Mterfd1 |
Location: |
Chr.13:66905456-66933072 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mterf3 | NM_025547.3 | 5 | 899 | NP_079823.2 |
AK002299.1 | 5 | 840 | ||
AK010525.1 | 5 | 894 | BAB27006.1 | |
AK011934.1 | 5 | 887 | BAB27924.3 | |
BC025173.1 | 5 | 868 | AAH25173.1 | |
BC085282.1 | 5 | 869 | AAH85282.1 |
