Assay Details
Target Gene Details
Entrez Gene ID: | 75512 |
Gene Name: | glutathione peroxidase 6 |
Gene Aliases: |
1700020G18Rik, Ry2d1, olfactory |
Location: |
Chr.13:21295368-21319625 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx6 | NM_145451.3 | NP_663426.2 | ||
| BC013526.1 | AAH13526.1 | |||
| CB955421.1 | 1 | 53 |
More Information
Set Membership: |
|

Genomic Map