Assay Details
Target Gene Details
Entrez Gene ID: | 110521 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 1 |
Gene Aliases: |
Cryabp1 |
Location: |
Chr.13:42051969-42185030 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hivep1 | NM_007772.2 | 2 | 127 | NP_031798.2 |
AK039808.1 | 2 | 127 | BAE20554.1 |
