Assay Details
Target Gene Details
Entrez Gene ID: | 17828 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 5, muted |
Gene Aliases: |
1810074A19Rik, Muted, mu |
Location: |
Chr.13:38602706-38635884 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s5 | NM_139063.1 | NP_620702.1 | ||
| AF426433.1 | AAL99384.1 | |||
| AK039050.1 | BAC30220.1 | |||
| AK075808.1 | BAC35976.1 | |||
| AK170614.1 | BAE41913.1 | |||
| BC023184.1 | AAH23184.1 | |||
| BC024720.1 | AAH24720.1 |

Genomic Map